Kanamycin sulfate
Kanamycin sulfate
Kanamycin sulfate
Thermo Scientific Chemicals

Kanamycin sulfate

Kanamycin sulfate, CAS # 25389-94-0, is an aminoglycoside sulfate salt with antibacterial activity.
Have Questions?
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 450811000
Price (USD)
914.00
Each
Request bulk or custom format
Price (USD)
914.00
Each
Chemical Identifiers
CAS25389-94-0
IUPAC Name(2R,3S,4S,5R,6R)-2-(aminomethyl)-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-3-{[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}oxane-3,4,5-triol; sulfuric acid
Molecular FormulaC18H38N4O15S
InChI KeyOOYGSFOGFJDDHP-KMCOLRRFSA-N
SMILESOS(O)(=O)=O.NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Loss on drying=<4.0 %
pH6.5 to 8.5 (1 % in water)
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Kanamycin sulfate is the sulfate salt form of kanamycin A
  • It was isolated from Streptomyces kanamyceticus
  • This compound inhibits translocation during protein biosynthesis. It binds to the 30S ribosomal subunit, causing misreading of the mRNA sequence

Application

  • Kanamycin sulfate shows antibacterial activity. It is effective against gram-positive and gram-negative bacteria, as well as Mycoplasma species
  • This compound is also involved in translating prokaryotic ribosomes
  • In molecular biology experiments, it is commonly used to select and isolate transformants expressing the kanamycin resistance gene, and plants that have the NPT II (APH3) gene incorporated
RUO – Research Use Only