Sodium sulfate decahydrate, 99+%, for analysis
Sodium sulfate decahydrate, 99+%, for analysis
Sodium sulfate decahydrate, 99+%, for analysis
Thermo Scientific Chemicals

Sodium sulfate decahydrate, 99+%, for analysis

CAS: 7727-73-3 ∣ Na2O4S·10H2O ∣ 322.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 451100010
Price (USD)
45.50
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
45.50
Each
Chemical Identifiers
CAS7727-73-3
IUPAC Namedisodium decahydrate sulfate
Molecular FormulaH20Na2O14S
InChI KeyRSIJVJUOQBWMIM-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration after Ion exchange>=99.0 %
pH5.2 to 9.2 (5% soln. at 25°C)
Arsenic (As)=<0.5 ppm
Calcium (Ca)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only