3-Aminooxetane, 95%, Thermo Scientific Chemicals
3-Aminooxetane, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Aminooxetane, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
250 mg
Catalog number 451215000
Price (USD)/ Each
253.00
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
253.00
Add to cart
3-Aminooxetane, 95%, Thermo Scientific Chemicals
Catalog number451215000
Price (USD)/ Each
253.00
-
Add to cart
Chemical Identifiers
IUPAC Nameoxetan-3-amine
Molecular FormulaC3H7NO
InChI KeyOJEOJUQOECNDND-UHFFFAOYSA-N
SMILESNC1COC1
Molecular Weight (g/mol)73.10
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94.0 %
Appearance (Form)Liquid
Appearance (Color)Colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only