4,4,5,5-Tetramethyl-2-(2-methyl-3-furyl)-1,3,2-dioxaborolane, 90%
4,4,5,5-Tetramethyl-2-(2-methyl-3-furyl)-1,3,2-dioxaborolane, 90%
4,4,5,5-Tetramethyl-2-(2-methyl-3-furyl)-1,3,2-dioxaborolane, 90%
Thermo Scientific Chemicals

4,4,5,5-Tetramethyl-2-(2-methyl-3-furyl)-1,3,2-dioxaborolane, 90%

CAS: 864776-02-3 | C11H17BO3 | 208.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 451520010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS864776-02-3
IUPAC Name4,4,5,5-tetramethyl-2-(2-methylfuran-3-yl)-1,3,2-dioxaborolane
Molecular FormulaC11H17BO3
InChI KeyJMLBPHNESOKSEV-UHFFFAOYSA-N
SMILESCC1=C(C=CO1)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=89.0 %
Appearance (Color)Colorless to orange
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only