Tin(IV) oxide, 99%, pure
Tin(IV) oxide, 99%, pure
Tin(IV) oxide, 99%, pure
Thermo Scientific Chemicals

Tin(IV) oxide, 99%, pure

CAS: 18282-10-5 | O2Sn | 150.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
Catalog number 451811000
Price (USD)
30.65
Online Exclusive
34.10
Save 3.45 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
30.65
Online Exclusive
34.10
Save 3.45 (10%)
Each
Chemical Identifiers
CAS18282-10-5
IUPAC Namestannanedione
Molecular FormulaO2Sn
InChI KeyXOLBLPGZBRYERU-UHFFFAOYSA-N
SMILESO=[Sn]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Purity>=98.5 % (metals basis)
Appearance (Color)White to light grey
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only