Zinc acetate dihydrate, 97%, specified according to the requirements of USP
Zinc acetate dihydrate, 97%, specified according to the requirements of USP
Zinc acetate dihydrate, 97%, specified according to the requirements of USP
Thermo Scientific Chemicals

Zinc acetate dihydrate, 97%, specified according to the requirements of USP

CAS: 5970-45-6 | C4H6O4Zn·2H2O | 219.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 451880010
Price (USD)
79.90
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
79.90
Each
Chemical Identifiers
CAS5970-45-6
IUPAC Namezinc(2+) diacetate dihydrate
Molecular FormulaC4H10O6Zn
InChI KeyBEAZKUGSCHFXIQ-UHFFFAOYSA-L
SMILESO.O.[Zn++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration Complexometric98.0 to 102.0 %
pH6.0 to 8.0 (5 % solution)
Chloride (Cl)=<0.005 %
Sulfate (SO4)=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only