Toluene, technical
Toluene, technical
Toluene, technical
Thermo Scientific Chemicals

Toluene, technical

CAS: 108-88-3 | C7H8 | 92.14 g/mol
Catalog NumberQuantityPackaging
4519700505 LPlastic Drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 451970050
Price (JPY)
-
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS108-88-3
IUPAC Nametoluene
Molecular FormulaC7H8
InChI KeyYXFVVABEGXRONW-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4900 to 1.5030 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Density0.865 to 0.872 kg/l
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only