Ascites Conditioning Reagent for Melon™ Gel Monoclonal IgG Purification Kit
Thermo Scientific Ascites Conditioning Reagent, formulation: proprietary solution sufficient for treating 125 mL of ascites fluid for optimal performance inRead more
Have Questions?
Catalog Number
Quantity
45219
5 mL
Catalog number 45219
Price (USD)
74.75
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
74.75
Each
Thermo Scientific Ascites Conditioning Reagent, formulation: proprietary solution sufficient for treating 125 mL of ascites fluid for optimal performance in Melon Gel purification.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce Ascites Conditioning Reagent
Quantity5 mL
Reagent TypeConditioning Reagent
FormatKit
Product TypeAscites Conditioning Reagent
Unit SizeEach
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Frequently asked questions (FAQs)
Do you offer a kit for antibody purification?
For purification of antibodies, we offer Melon Gel, which binds other proteins from serum, ascites or culture supernatant, allowing the IgG to be collected in the flow-through. As low pH is typically used to elute IgG from Protein A, G, A/G or L, the use of Melon Gel can spare the IgG from exposure to such harsh conditions, which can be particularly important for monoclonal antibodies.