Nunc™ MicroWell™ MiniTrays
Nunc™ MicroWell™ MiniTrays
Nunc™ MicroWell™ MiniTrays
Nunc™ MicroWell™ MiniTrays
Nunc™ MicroWell™ MiniTrays
Nunc™ MicroWell™ MiniTrays
Thermo Scientific™

Nunc™ MicroWell™ MiniTrays

Create more space in lab freezers and inubators with Nunc MicroWell MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWell CountClosure IncludedSterility
45225660YesSterile
43873372YesNon-sterile
43922560YesNon-sterile
47037872YesNon-sterile
47240060NoSterile
Catalog number 452256
Price (USD)
1,515.67
Online Exclusive
1,768.13
Save 252.46 (14%)
Case of 480
Well Count:
60
Closure Included:
Yes
Sterility:
Sterile
Price (USD)
1,515.67
Online Exclusive
1,768.13
Save 252.46 (14%)
Case of 480

Create more space in lab freezers and inubators with Thermo Scientific™ Nunc™ MicroWell™ MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.

  • Limited adsorption of proteins
  • Lids have keyed tabs for proper orientation
  • Large raised alphanumeric coordinates
  • Unique surface treatment to maximize sample and reagent mixing
  • MiniTrays for serological applications have slightly smaller well dimensions
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Closure IncludedYes
Closure MaterialPolystyrene
IncludesLid
Length (Metric)84 mm
No. per Case480
No. per Pack120
ShapeRectangular
TypeCell Culture Dish
Width (Metric)59 mm
Bottom ShapeFlat
Height (Metric)59 mm
MaterialPolystyrene
SterilitySterile
Surface TreatmentNunclon Delta
Well Count60
Unit SizeCase of 480