(Isocyanoimino)triphenylphosphorane, 90%
(Isocyanoimino)triphenylphosphorane, 90%
(Isocyanoimino)triphenylphosphorane, 90%
Thermo Scientific Chemicals

(Isocyanoimino)triphenylphosphorane, 90%

CAS: 73789-56-7 | C19H15N2P | 302.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 452920010
Price (USD)
64.65
Special offer
Online exclusive
Ends: 28-Dec-2026
75.80
Save 11.15
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
64.65
Special offer
Online exclusive
Ends: 28-Dec-2026
75.80
Save 11.15
Each
Chemical Identifiers
CAS73789-56-7
IUPAC Nameisocyano(triphenyl-λ⁵-phosphanylidene)amine
Molecular FormulaC19H15N2P
InChI KeyNIDTXBFHPXMXTR-UHFFFAOYSA-N
SMILES[C-]#[N+]N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=88 %
Appearance (Color)White to light yellow-orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only