Stachyose hydrate, technical
Stachyose hydrate, technical
Stachyose hydrate, technical
Thermo Scientific Chemicals

Stachyose hydrate, technical

CAS: 54261-98-2 | C24H42O21 | 666.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 453140010
Price (USD)
195.65
Special offer
Online exclusive
Ends: 28-Dec-2026
230.00
Save 34.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
195.65
Special offer
Online exclusive
Ends: 28-Dec-2026
230.00
Save 34.35
Each
Chemical Identifiers
CAS54261-98-2
IUPAC Name(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-({[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-({[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}methyl)oxane-3,4,5-triol
Molecular FormulaC24H42O21
InChI KeyUQZIYBXSHAGNOE-CYDMKDQUNA-N
SMILESOC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO[C@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light beige
Appearance (Form)Powder
Infrared spectrumConforms
Water=<12 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only