Graphene, nanoplatelets 2-10 nm
Graphene, nanoplatelets 2-10 nm
Graphene, nanoplatelets 2-10 nm
Thermo Scientific Chemicals

Graphene, nanoplatelets 2-10 nm

CAS: 1034343-98-0 | Cx | 16.04 g/mol
Have Questions?
Catalog NumberQuantity
45316050050 g
Catalog number 453160500
Price (USD)
310.65
Online exclusive
345.00
Save 34.35
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
310.65
Online exclusive
345.00
Save 34.35
Each
Chemical Identifiers
CAS1034343-98-0
IUPAC NameGraphene
Molecular FormulaCx
SMILESC
Synonymmethane collocarb carbene, 13c-labeled 13C-labeled carbene kosmos spheron magecol grosafe humenegro furnal
View more
SpecificationsSpecification SheetSpecification Sheet
Thickness2 to 10 nm
Particle size(Diameter) approx. 5 µm
Strength5 Gpa (Tensile strength)
Thickness2 to 10 nm
Particle size(Diameter) approx. 5 µm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only