Lead(II) acetate basic, ACS reagent
Lead(II) acetate basic, ACS reagent
Lead(II) acetate basic, ACS reagent
Thermo Scientific Chemicals

Lead(II) acetate basic, ACS reagent

CAS: 51404-69-4 | C4H10O8Pb3 | 807.69 g/mol
Have Questions?
Catalog NumberQuantity
4532200101 kg
Catalog number 453220010
Price (USD)
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS51404-69-4
IUPAC Name(acetyloxy)plumbyl acetate dihydrate lead
Molecular FormulaC4H10O6Pb2
InChI KeyXAOPZCQFBUCRKR-UHFFFAOYSA-L
SMILESO.O.[Pb].CC(=O)O[Pb]OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<0.003 %
Nitrate (NO3)=<0.003 %
Calcium (Ca)=<0.01 %
Copper (Cu)=<0.002 %
Iron (Fe)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only