2,4-Dibromopyridine, 97%
2,4-Dibromopyridine, 97%
2,4-Dibromopyridine, 97%
Thermo Scientific Chemicals

2,4-Dibromopyridine, 97%

CAS: 58530-53-3 | C5H3Br2N | 236.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 453700010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS58530-53-3
IUPAC Name2,4-dibromopyridine
Molecular FormulaC5H3Br2N
InChI KeyPCMMSLVJMKQWMQ-UHFFFAOYSA-N
SMILESBrC1=CC=NC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow to brown
Appearance (Form)Low melting solid
Infrared spectrumConforms
Melting point35°C to 40°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only