L-Phenylalanine, 98.5-101.0%, specified according to the requirements of USP
L-Phenylalanine, 98.5-101.0%, specified according to the requirements of USP
L-Phenylalanine, 98.5-101.0%, specified according to the requirements of USP
Thermo Scientific Chemicals

L-Phenylalanine, 98.5-101.0%, specified according to the requirements of USP

CAS: 63-91-2 | C9H11NO2 | 165.19 g/mol
製品番号(カタログ番号) 453930250
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS63-91-2
IUPAC Name2-amino-3-phenylpropanoic acid
Molecular FormulaC9H11NO2
InChI KeyCOLNVLDHVKWLRT-UHFFFAOYNA-N
SMILESNC(CC1=CC=CC=C1)C(O)=O
さらに表示
Appearance (Color)Colorless to white
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration with HClO498.5 to 101.5 % (On dried base)
Sulfated ash=<0.4 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only