2-Bromo-1,3-thiazole-4-carboxylic acid, 98%
2-Bromo-1,3-thiazole-4-carboxylic acid, 98%
2-Bromo-1,3-thiazole-4-carboxylic acid, 98%
Thermo Scientific Chemicals

2-Bromo-1,3-thiazole-4-carboxylic acid, 98%

CAS: 5198-88-9 | C4H2BrNO2S | 208.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 454600010
Price (USD)
132.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
132.00
Each
Chemical Identifiers
CAS5198-88-9
IUPAC Name2-bromo-1,3-thiazole-4-carboxylate
Molecular FormulaC4HBrNO2S
InChI KeyBEGREHRAUWCAHV-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CSC(Br)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Color)White to light brown
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only