Quinoline-5-carboxylic acid, 97%
Quinoline-5-carboxylic acid, 97%
Quinoline-5-carboxylic acid, 97%
Thermo Scientific Chemicals

Quinoline-5-carboxylic acid, 97%

CAS: 7250-53-5 | C10H7NO2 | 173.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 454760050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7250-53-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white to light brown
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only