Sodium trifluoromethanesulfinate, 98%
Sodium trifluoromethanesulfinate, 98%
Sodium trifluoromethanesulfinate, 98%
Thermo Scientific Chemicals

Sodium trifluoromethanesulfinate, 98%

CAS: 2926-29-6 | CF3NaO2S | 156.06 g/mol
製品番号(カタログ番号) 454990050
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2926-29-6
IUPAC Namesodium trifluoromethanesulfinate
Molecular FormulaCF3NaO2S
InChI KeyKAVUKAXLXGRUCD-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)C(F)(F)F
さらに表示
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White to light yellow
Infrared spectrumConforms
Water=<1 %
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only