Sodium trifluoromethanesulfinate, 98%
Sodium trifluoromethanesulfinate, 98%
Sodium trifluoromethanesulfinate, 98%
Thermo Scientific Chemicals

Sodium trifluoromethanesulfinate, 98%

CAS: 2926-29-6 ∣ CF3NaO2S ∣ 156.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 454990250
Price (USD)
272.65
Special offer
Online exclusive
Ends: 28-Dec-2026
321.00
Save 48.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
272.65
Special offer
Online exclusive
Ends: 28-Dec-2026
321.00
Save 48.35
Each
Chemical Identifiers
CAS2926-29-6
IUPAC Namesodium trifluoromethanesulfinate
Molecular FormulaCF3NaO2S
InChI KeyKAVUKAXLXGRUCD-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White to light yellow
Infrared spectrumConforms
Water=<1 %
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only