Nalgene™ Rapid-Flow™ Sterile Filter Storage Bottles
Thermo Scientific™

Nalgene™ Rapid-Flow™ Sterile Filter Storage Bottles

Save time and minimize contamination with these radiation-sterilized filter storage bottles. Excellent as single-use storage containers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
455-10001 L
455-0500500 mL
455-0150150 mL
455-0250250 mL
Catalog number 455-1000
Price (EUR)
119,74
Online exclusive
135,00
Save 15,26 (11%)
Case of 12
Volume (Metric):
1 L
Price (EUR)
119,74
Online exclusive
135,00
Save 15,26 (11%)
Case of 12

Save time and minimize contamination with gamma-sterilized Thermo Scientific™ Nalgene™ Rapid-Flow™ Filter Storage Bottles. Use with Nalgene Bottle-Top filters or as replacement/extra receptacles for filter units. Excellent as single-use storage containers.

  • Headspace allows for extra serum or other additives to be added to the filtrate
  • Leakproof† screw caps to eliminate pH shift in bottle for longer life of stored media
  • Sterilized by gamma radiation and individually bagged for a 5-year sterile shelf life
  • Non-pyrogenic and non-cytotoxic
  • Available with Fit for Purpose Sterile documentation

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Cap MaterialVirgin HDPE
Closure ColorWhite
ModelWide
MouthWide
No. per Case12
No. per Pack12
Volume (Metric)1 L
Fit for PurposeSterile
MaterialVirgin Polystyrene
SterilitySterile
Unit SizeCase of 12