Thermo Scientific Chemicals

Ceftriaxone sodium salt hemiheptahydrate

Catalog number: 455420050
5 g, Each
Thermo Scientific Chemicals

Ceftriaxone sodium salt hemiheptahydrate

Catalog number: 455420050
5 g, Each
Quantity

Chemical Identifiers

CAS
104376-79-6
IUPAC Name
tetrasodium 3-({[(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-2-carboxylato-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}sulfanyl)-2-methyl-5,6-dioxo-1,2,5,6-tetrahydro-1,2,4-triazin-1-ide (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-{[(2-methyl-6-oxido-5-oxo-2,5-dihydro-1,2,4-triazin-3-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate heptahydrate
Molecular Formula
C36H46N16Na4O21S6
InChI Key
RGDBKADCOSIOEV-MAODNAKNSA-J
SMILES
O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].CON=C(/C(=O)N[C@H]1[C@H]2SCC(CSC3=NC(=O)C(=O)[N-]N3C)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1.CON=C(/C(=O)N[C@H]1[C@H]2SCC(CSC3=NC(=O)C([O-])=NN3C)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Water
=<10 %
Appearance (Form)
Crystalline powder
HPLC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Ceftriaxone is the sodium salt of ceftriaxone, a beta-lactam antibiotic of the cephalosporin class that works by inactivating penicillin-binding proteins

Applications

• Penicillin-binding protein inactivation inhibits peptidoglycan cross-linkage and leads to destruction of the cell wall and subsequent lysis

• It exhibits antibiotic activity against gram-negative bacteria

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text