3-(Bromoacetyl)pyridine hydrobromide, 98%
3-(Bromoacetyl)pyridine hydrobromide, 98%
3-(Bromoacetyl)pyridine hydrobromide, 98%
Thermo Scientific Chemicals

3-(Bromoacetyl)pyridine hydrobromide, 98%

CAS: 17694-68-7 | C7H7Br2NO | 280.95 g/mol
Quantity:
5 g
25 g
Catalog number 455520250
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS17694-68-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Argentometric>=97.5 %
Melting point188°C to 197°C
Appearance (Color)White to yellow to beige
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only