4-tert-Butylcyclohexylamine, 97%, mixture of cis and trans
4-tert-Butylcyclohexylamine, 97%, mixture of cis and trans
4-tert-Butylcyclohexylamine, 97%, mixture of cis and trans
Thermo Scientific Chemicals

4-tert-Butylcyclohexylamine, 97%, mixture of cis and trans

CAS: 5400-88-4 | C10H21N | 155.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Catalog number 455600250
Price (USD)
139.00
Each
Quantity:
25 mL
Request bulk or custom format
Price (USD)
139.00
Each
Chemical Identifiers
CAS5400-88-4
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4630 to 1.4660 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=96.0 % (total of isomers)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only