Arbutin, 98%
Arbutin, 98%
Arbutin, 98%
Thermo Scientific Chemicals

Arbutin, 98%

CAS: 497-76-7 | C12H16O7 | 272.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 455640250
Price (EUR)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS497-76-7
IUPAC Name2-(hydroxymethyl)-6-(4-hydroxyphenoxy)oxane-3,4,5-triol
Molecular FormulaC12H16O7
InChI KeyBJRNKVDFDLYUGJ-UHFFFAOYNA-N
SMILESOCC1OC(OC2=CC=C(O)C=C2)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Loss on drying=<2 %
Melting point196°C to 201°C
Specific optical rotation-63° to -67° (20°C, 589 nm) (c=3, H2O)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only