4-Hydroxyphenyl benzoate, 98%
4-Hydroxyphenyl benzoate, 98%
4-Hydroxyphenyl benzoate, 98%
Thermo Scientific Chemicals

4-Hydroxyphenyl benzoate, 98%

CAS: 2444-19-1 | C13H10O3 | 214.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 455730250
Price (EUR)
138,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
138,00
Each
Chemical Identifiers
CAS2444-19-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Melting point162°C to 166°C
Appearance (Form)Crystalline powder
Loss on drying=<0.5 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only