Effectively centrifugalize samples during pelleting and gradient separations with the Thermo Scientific™ S80-AT3 Fixed Angle Rotor, offering 8 x 8.3 capacity at speeds up to 80,000rpm.
Features:
Capacity: 8 × 8.3mL
Max. Speed/RCF: 80,000rpm/414,630 x g
K factor: 23.2
Compatible with:
Thermo Scientific™ Sorvall™ MTX 150 Series Micro-Ultracentrifuge, MX Series Floor Model Micro-Ultracentrifuge
Recommended for:
Pelleting and gradient separations.
Specifications
Angle30°
Avg. RCF323,126 x g
DescriptionS80-AT3 Fixed Angle Rotor
For Use With (Equipment)Thermo Scientific™ Sorvall™ MTX and MX Plus micro-ultraCentrifuges