1,4-Cineole, 85%
1,4-Cineole, 85%
1,4-Cineole, 85%
Thermo Scientific Chemicals

1,4-Cineole, 85%

CAS: 470-67-7 | C10H18O | 154.25 g/mol
製品番号(カタログ番号) 456061000
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS470-67-7
IUPAC Name1-methyl-4-(propan-2-yl)-7-oxabicyclo[2.2.1]heptane
Molecular FormulaC10H18O
InChI KeyRFFOTVCVTJUTAD-UHFFFAOYSA-N
SMILESCC(C)C12CCC(C)(CC1)O2
さらに表示
Infrared spectrumConforms
GC>=80 %
Appearance (Color)Clear colorless
Refractive index1.4450 to 1.4560 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only