1-Phenyl-2-propyn-1-one, 97%
1-Phenyl-2-propyn-1-one, 97%
1-Phenyl-2-propyn-1-one, 97%
Thermo Scientific Chemicals

1-Phenyl-2-propyn-1-one, 97%

CAS: 3623-15-2 | C9H6O | 130.15 g/mol
Have Questions?
Catalog NumberQuantity
4564300101 g
Catalog number 456430010
Price (USD)
308.65
Special offer
Online exclusive
Ends: 28-Dec-2026
363.00
Save 54.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
308.65
Special offer
Online exclusive
Ends: 28-Dec-2026
363.00
Save 54.35
Each
Chemical Identifiers
CAS3623-15-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystalline powder
GC>=96.0 %
Appearance (Color)White to yellow
Infrared spectrumConforms
Melting point48°C to 53°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only