Ciprofloxacin hydrochloride hydrate, 98%
Ciprofloxacin hydrochloride hydrate, 98%
Ciprofloxacin hydrochloride hydrate, 98%
Thermo Scientific Chemicals

Ciprofloxacin hydrochloride hydrate, 98%

Ciprofloxacin hydrochloride hydrate, CAS # 128074-72-6, is the hydrate form of the hydrochloride salt of ciprofloxacin, a broad-spectrum antibiotic.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
45688025025 g
4568800505 g
Catalog number 456880250
Price (USD)
151.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
151.00
Each
Chemical Identifiers
CAS128074-72-6
IUPAC Name1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride
Molecular FormulaC17H19ClFN3O3
InChI KeyDIOIOSKKIYDRIQ-UHFFFAOYSA-N
SMILESCl.OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Water4.0 to 7.0 %
Residue after ignition=<0.1 %
Infrared spectrumConforms
Appearance (Color)White to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Ciprofloxacin HCl is a fluoroquinolone antibiotic that works by binding to bacterial DNA gyrase/DNA topoisomerase, thus inhibiting DNA replication and subsequent bacterial cell division
• It can cause double-stranded breaks in the bacterial chromosome
• Ciprofloxacin HCl is an analog of the antibiotic nalidixic acid
• Works as an antibiotic against both gram-positive and gram-negative bacteria and can have fungicidal activity
Applications
• Used for elimination of mycoplasma in cell culture
• Can be used to study antimicrobial activity and resistance in laboratory settings
RUO – Research Use Only
Literature references
Schmitt, K.; Däubener, W.; Bitter-Suermann, D.; Hadding, U. A safe and efficient method for elimination of cell culture mycoplasmas using ciprofloxacin. J Immunol Methods. 1988, 109(1), 17-25.
Jomehzadeh, N.; Saki, M.; Ahmadi, K.; Zandi, G. The prevalence of plasmid-mediated quinolone resistance genes among Escherichia coli strains isolated from urinary tract infections in southwest Iran. Mol Biol Rep. 2022.
D.C. Hooper, et al. Mechanisms of action of and resistance to ciprofloxacin. Am. J. Med. 1987, 82, (4A), 12-20.