Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™
Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™
Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™
Thermo Scientific Chemicals

Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™

CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Catalog number 457070010
Price (USD)
134.00
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
134.00
Each
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Peroxides=<0.0075 %
Residue after evaporation=<0.001 %
Appearance (Form)Clear liquid
GC>=99.80 %
Water=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only