Copper(II) pyrithione
Copper(II) pyrithione
Copper(II) pyrithione
Thermo Scientific Chemicals

Copper(II) pyrithione

CAS: 14915-37-8 | C10H8CuN2O2S2 | 315.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 457210050
Price (EUR)
322,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
322,00
Each
Chemical Identifiers
CAS14915-37-8
IUPAC Namecopper(2+) bis((1-oxidopyridin-1-ium-2-yl)sulfanide)
Molecular FormulaC10H8CuN2O2S2
InChI KeyFNQNNKLJHNPTGT-UHFFFAOYSA-L
SMILES[Cu++].[O-][N+]1=CC=CC=C1[S-].[O-][N+]1=CC=CC=C1[S-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only