2-Methyl-4-nitrobenzoic acid, 98%
2-Methyl-4-nitrobenzoic acid, 98%
2-Methyl-4-nitrobenzoic acid, 98%
Thermo Scientific Chemicals

2-Methyl-4-nitrobenzoic acid, 98%

CAS: 1975-51-5 | C8H7NO4 | 181.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 457810250
Price (EUR)
129,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
129,00
Each
Chemical Identifiers
CAS1975-51-5
IUPAC Name2-methyl-4-nitrobenzoic acid
Molecular FormulaC8H7NO4
InChI KeyXXXOBNJIIZQSPT-UHFFFAOYSA-N
SMILESCC1=CC(=CC=C1C(O)=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only