Evodiamine, 98%
Evodiamine, 98%
Evodiamine, 98%
Thermo Scientific Chemicals

Evodiamine, 98%

CAS: 518-17-2 | C19H17N3O | 303.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 457890010
Price (EUR)
64,20
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
64,20
Each
Chemical Identifiers
CAS518-17-2
IUPAC Name(1S)-21-methyl-3,13,21-triazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8,15,17,19-heptaen-14-one
Molecular FormulaC19H17N3O
InChI KeyTXDUTHBFYKGSAH-SFHVURJKSA-N
SMILESCN1[C@H]2N(CCC3=C2NC2=CC=CC=C32)C(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Color)White to cream
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only