Cyclosporin A, 98%
Cyclosporin A, 98%
Cyclosporin A, 98%
Thermo Scientific Chemicals

Cyclosporin A, 98%

Cyclosporin A, 98%, CAS # 59865-13-3, is a compound that shows immunosuppressive, antifungal, antirheumatic, and dermatologic activities. It is also a calcineurin and enzyme inhibitor. | CAS: 59865-13-3 | C62H111N11O12 | 1202.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 457970050
Price (USD)
485.65
Special offer
Online exclusive
Ends: 28-Dec-2026
571.00
Save 85.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
485.65
Special offer
Online exclusive
Ends: 28-Dec-2026
571.00
Save 85.35
Each
Chemical Identifiers
CAS59865-13-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
Loss on drying=<2.0 % (105°C, 3 hrs)
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Cyclosporin A is a non-polar cyclic oligopeptide produced by the fungus Tolypocladium inflatum

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Staudinger, L. A.; Spano, S. J.; Lee, W. S.; Coelho, N.; Moriarty, T. J.; McCulloch, C. A. Role of discoidin domain receptor 1 in dysregulation of collagen remodeling by cyclosporin A. Int. J. Biochem. Cell Biol. 2015, 62, 80-87.
  2. Petrillo, C.; Cesana, D.; Piras, F.; Bartolaccini, S.; Naldini, L.; Montini, E.; Rudnitski, A. K. Cyclosporin A and Rapamycin Relieve Distinct Lentiviral Restriction Blocks in Hematopoietic Stem and Progenitor Cells. Molecular Therapy 2015, 23 (2), 352-362.

Frequently asked questions (FAQs)

Which solvents can I use to dissolve cyclosporin A, 98%?

Cyclosporin A is soluble in ethanol, chloroform, methylene chloride/dichloromethane, and DMSO.