Poly(vinyl alcohol), 98% hydrolyzed, approx. M.W. 13,000
Poly(vinyl alcohol), 98% hydrolyzed, approx. M.W. 13,000
Poly(vinyl alcohol), 98% hydrolyzed, approx. M.W. 13,000
Thermo Scientific Chemicals

Poly(vinyl alcohol), 98% hydrolyzed, approx. M.W. 13,000

CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 458240250
Price (USD)
44.60
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
44.60
Each
Chemical Identifiers
CAS9002-89-5
Molecular Formula(C2H4O)n
SMILESOC(-*)C-*
MDL NumberMFCD00081922
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Volatile matter=<5 %
Ash=<1.2 % (as Na2O)
Appearance (Color)Off-white to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only