Thermo Scientific™

2'-Deoxycytidine hydrochloride, 98%, Thermo Scientific™

25g, Each
Thermo Scientific™

2'-Deoxycytidine hydrochloride, 98%, Thermo Scientific™

25g, Each
Quantity

Chemical Identifiers

CAS
3992-42-5
Appearance (Color)
Off-white to light yellow
Infrared spectrum
Conforms
HPLC
>=97.5 %
Water
=<8 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only

Figures