Omeprazole, 98+%
Omeprazole, 98+%
Omeprazole, 98+%
Thermo Scientific Chemicals

Omeprazole, 98+%

CAS: 73590-58-6 | C17H19N3O3S | 345.42 g/mol
製品番号(カタログ番号) 458520050
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS73590-58-6
IUPAC Name6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methanesulfinyl]-1H-1,3-benzodiazole
Molecular FormulaC17H19N3O3S
InChI KeySUBDBMMJDZJVOS-UHFFFAOYNA-N
SMILESCOC1=CC=C2N=C(NC2=C1)S(=O)CC1=NC=C(C)C(OC)=C1C
さらに表示
Appearance (Color)White to off-white
Appearance (Form)Powder
Loss on drying=<0.5 % (60°C, 4 h) (vacuum)
Infrared spectrumConforms
HPLC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only