Trimethylsilanol
Trimethylsilanol
Trimethylsilanol
Thermo Scientific Chemicals

Trimethylsilanol

CAS: 1066-40-6 | C3H9LiOSi | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 458570050
Price (USD)
59.65
Online Exclusive
65.80
Save 6.15 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
59.65
Online Exclusive
65.80
Save 6.15 (9%)
Each
Chemical Identifiers
CAS1066-40-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only