Ammonium hydroxide, extra pure, 10 wt% in water
Ammonium hydroxide, extra pure, 10 wt% in water
Ammonium hydroxide, extra pure, 10 wt% in water
Thermo Scientific Chemicals

Ammonium hydroxide, extra pure, 10 wt% in water

CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number 458680010
Price (USD)
32.00
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
32.00
Each
Chemical Identifiers
CAS1336-21-6
IUPAC Nameamine hydrate
Molecular FormulaH5NO
InChI KeyVHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILESN.O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear solution
Color scale=<10 APHA
Strength9.5 to 10.5 %
Residue after evaporation=<0.0025 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only