Simvastatin, 98%
Simvastatin, 98%
Simvastatin, 98%
Thermo Scientific Chemicals

Simvastatin, 98%

CAS: 79902-63-9 | C25H38O5 | 418.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 458840050
Price (USD)
141.65
Special offer
Online exclusive
Ends: 28-Dec-2026
167.00
Save 25.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
141.65
Special offer
Online exclusive
Ends: 28-Dec-2026
167.00
Save 25.35
Each
Chemical Identifiers
CAS79902-63-9
IUPAC Name(1S,3R,7S,8S,8aR)-8-{2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl}-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2,2-dimethylbutanoate
Molecular FormulaC25H38O5
InChI KeyRYMZZMVNJRMUDD-HGQWONQESA-N
SMILESCCC(C)(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point125°C to 135°C
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only