Cytisine, 98%
Cytisine, 98%
Cytisine, 98%
Thermo Scientific Chemicals

Cytisine, 98%

CAS: 485-35-8 | C11H14N2O | 190.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 458852500
Price (USD)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS485-35-8
IUPAC Name(1S,9S)-7,11-diazatricyclo[7.3.1.0²,⁷]trideca-2,4-dien-6-one
Molecular FormulaC11H14N2O
InChI KeyANJTVLIZGCUXLD-IUCAKERBSA-N
SMILESO=C1C=CC=C2[C@@H]3CNC[C@H](C3)CN12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=97.5 %
Appearance (Color)White to yellow
Infrared spectrumConforms
Loss on drying=<5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only