Methyl 4-fluorobenzoate, 98%
Methyl 4-fluorobenzoate, 98%
Methyl 4-fluorobenzoate, 98%
Thermo Scientific Chemicals

Methyl 4-fluorobenzoate, 98%

CAS: 403-33-8 | C8H7FO2 | 154.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 459241000
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS403-33-8
IUPAC Namemethyl 4-fluorobenzoate
Molecular FormulaC8H7FO2
InChI KeyMSEBQGULDWDIRW-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4930 to 1.4950 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Colorless to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only