Meloxicam, 99-101%
Meloxicam, 99-101%
Meloxicam, 99-101%
Thermo Scientific Chemicals

Meloxicam, 99-101%

CAS: 71125-38-7 | C14H13N3O4S2 | 351.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 459550050
Price (USD)
58.60
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
58.60
Each
Chemical Identifiers
CAS71125-38-7
IUPAC Name(3E)-3-{hydroxy[(5-methyl-1,3-thiazol-2-yl)amino]methylidene}-2-methyl-3,4-dihydro-2H-1λ⁶,2-benzothiazine-1,1,4-trione
Molecular FormulaC14H13N3O4S2
InChI KeyDWMREKMVXIFPFM-ACCUITESSA-N
SMILESCN1\C(=C(\O)NC2=NC=C(C)S2)C(=O)C2=CC=CC=C2S1(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<0.5 % (105°C, 4 hr)
Titration with HClO499.0 to 101.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only