S140-AT Fixed Angle Rotor for MTX and MX Plus Micro-Ultracentrifuges
S140-AT Fixed Angle Rotor for MTX and MX Plus Micro-Ultracentrifuges
S140-AT Fixed Angle Rotor for MTX and MX Plus Micro-Ultracentrifuges
Thermo Scientific™

S140-AT Fixed Angle Rotor for MTX and MX Plus Micro-Ultracentrifuges

Support a variety of applications with this rotor, which operates at a maximum of 1,048,000 x g.
Have Questions?
Catalog NumberCapacity
4597810 x 2 mL
Catalog number 45978
Price (USD)
25,100.00
Each
Capacity:
10 x 2 mL
Price (USD)
25,100.00
Each
Support applications that require extreme force with the Thermo Scientific™ S140-AT Fixed Angle Rotor; this rotor is capable of operating at over 1,048,000 x g.

Features:

  • Fixed angle (35°) titanium rotor
  • Capacity: 10 × 2mL
  • Maximum Speed/RCF: 140,000rpm/1,048,000 x g
  • K factor: 5

Compatible with:

Thermo Scientific™ Sorvall™ MTX 150 Series Micro-Ultracentrifuge, MX Series Floor Model Micro-Ultracentrifuge

Recommended for:

For rapid separation of serum lipoproteins, DNA, RNA and subcellular fractionation.

Specifications
Angle35°
DescriptionS140-AT Fixed Angle Rotor
For Use With (Equipment)Thermo Scientific™ Sorvall™ MTX and MX Plus micro-ultraCentrifuges
K Factor5
Max. Tube Diameter11 mm
Max. Tube Length34 mm
Min. RCF713,718 x g
Net Weight1.8 lb. (0.8 kg)
Radius (Metric)47.9 mm
Radius (Metric) Min.32.6 mm
Capacity10 x 2 mL
MaterialTitanium
Max. RCF1,048,684 x g
Max. Speed140,000 rpm
TypeFixed Angle Rotor
Unit SizeEach