2-Chloro-3-methoxybenzoic acid, 97%
2-Chloro-3-methoxybenzoic acid, 97%
2-Chloro-3-methoxybenzoic acid, 97%
Thermo Scientific Chemicals

2-Chloro-3-methoxybenzoic acid, 97%

CAS: 33234-36-5 | C8H7ClO3 | 186.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 459900050
Price (USD)
211.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
211.00
Each
Chemical Identifiers
CAS33234-36-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder or crystalline powder or needles
Infrared spectrumConforms
Melting point160°C to 165°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only