CD1a Monoclonal Antibody (HI149), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD1a Monoclonal Antibody (HI149), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD1a Monoclonal Antibody (HI149), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0019-42)