CD8a Monoclonal Antibody (OX8), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD8a Monoclonal Antibody (OX8), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (OX8), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0084-82, 46-0084-80)