CD10 Monoclonal Antibody (SN5c), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD10 Monoclonal Antibody (SN5c), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD10 Monoclonal Antibody (SN5c), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0108-41, 46-0108-42)