CD11b/c Monoclonal Antibody (OX42), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD11b/c Monoclonal Antibody (OX42), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD11b/c Monoclonal Antibody (OX42), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0110-80, 46-0110-82)