CD11a (LFA-1alpha) Monoclonal Antibody (M17/4), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD11a (LFA-1alpha) Monoclonal Antibody (M17/4), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD11a (LFA-1alpha) Monoclonal Antibody (M17/4), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0111-80, 46-0111-82)